Succinic anhydride 97%

Supplier: Ambeed
Danger

Synonyms: Dihydrofuran-2,5-dione

A155638-100G A155638-25G
76783-558EA 14 USD
76783-558 76783-560
Succinic anhydride 97%
Succinic anhydride

Precaución: for Research Use Only

Formula: C₄H₄O₃
MDL Number: MFCD00005525
CAS Number: 108-30-5
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR