2-Amino-N,N-dimethylbenzenesulfonamide 98%

Supplier: Ambeed
Warning

A155608-100MG A155608-1G A155608-250MG A155608-5G
76783-550EA 6 USD
76783-550 76783-552 76783-554 76783-556
2-Amino-N,N-dimethylbenzenesulfonamide 98%
2-Amino-N,N-dimethylbenzenesulphonamide

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₂S
MDL Number: MFCD04035363
CAS Number: 54468-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR