Allylthiourea 98%

Supplier: Ambeed
Danger

Synonyms: N-Allylthiourea, Tiosinamine, 1-Allyl-2-thiourea, Allyl thiourea, Rhodalline, Thiosinamine, N-Allylthiourea

A154185-10G A154185-250MG A154185-25G A154185-5G
76783-526EA 8.49 USD
76783-526 76783-528 76783-530 76783-532
Allylthiourea 98%
Allylthiourea

Precaución: for Research Use Only

Formula: H₂CCHCH₂NHC(S)NH₂
MW: 116.19 g/mol
MDL Number: MFCD00004940
CAS Number: 109-57-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR