Saracatinib 98%

Supplier: Ambeed
Warning

Synonyms: AZD0530, N-(5-Chlorobenzo[d][1,3]dioxol-4-yl)-7-(2-(4-methylpiperazin-1-yl)ethoxy)-5-((tetrahydro-2H-pyran-4-yl)oxy)quinazolin-4-amine

A154045-10MG A154045-1MG A154045-50MG
76783-516EA 42 USD
76783-516 76783-518 77871-764
Saracatinib 98%
Saracatinib

Precaución: for Research Use Only

Formula: C₂₇H₃₂ClN₅O₅
MW: 542.03 g/mol
MDL Number: MFCD09832698
CAS Number: 379231-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR