AG-879 95%

Supplier: Ambeed
Warning

A153685-10MG A153685-1MG A153685-25MG A153685-50MG A153685-5MG A153685-100MG A153685-250MG
76783-488EA 21 USD
76783-488 76783-490 76783-492 76783-494 76783-496 77667-168 77871-761
AG-879 95%
AG-879

Precaución: for Research Use Only

Formula: C₁₈H₂₄N₂OS
MDL Number: MFCD00236450
CAS Number: 148741-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR