4-Bromo-2-iodobenzoic acid 97%

Supplier: Ambeed
Warning

A152612-100G A152612-10G A152612-1G A152612-25G A152612-5G
76783-430EA 195 USD
76783-430 76783-432 76783-434 76783-436 76783-438
4-Bromo-2-iodobenzoic acid 97%
4-Bromo-2-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄BrIO₂
MDL Number: MFCD12767500
CAS Number: 1133123-02-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR