N-Succinimidyl Acetate 98%

Supplier: Ambeed
Warning

Synonyms: N-Acetoxysuccinimide, Ac-Osu, Acetic Acid N-Succinimidyl Ester, 2,5-Dioxopyrrolidin-1-yl acetate

A152275-100G A152275-10G A152275-25G A152275-1G
76783-426EA 55 USD
76783-426 77549-916 77567-198 77600-168
N-Succinimidyl Acetate 98%
N-Succinimidyl Acetate

Precaución: For research use only.

Formula: C₆H₇NO₄
MW: 157.13 g/mol
MDL Number: MFCD00037916
CAS Number: 14464-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR