Tricaprylin 98%

Supplier: Ambeed
Warning

Synonyms: Propane-1,2,3-triyl trioctanoate, 1,2,3-Tri-n-octanoylglycerol, Tricaprylin, Tri-n-octanoin, Glycerol Tri-n-octanoate

A152150-100G A152150-25G A152150-5G
76783-408EA 85.94 USD
76783-408 76783-410 76783-412
Tricaprylin 98%
Tricaprylin

Precaución: for Research Use Only

Formula: C₂₇H₅₀O₆
MW: 470.69 g/mol
MDL Number: MFCD00036236
CAS Number: 538-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR