2-Fluoro-3-nitrobenzylalcohol 95%

Supplier: Ambeed
Warning

Synonyms: (2-Fluoro-3-nitrophenyl)methanol

A151196-1G A151196-250MG A151196-5G
76783-382EA 47 USD
76783-382 76783-384 76783-386
2-Fluoro-3-nitrobenzylalcohol 95%
2-Fluoro-3-nitrobenzylalkohol

Precaución: for Research Use Only

Formula: C₇H₆FNO₃
MDL Number: MFCD09835208
CAS Number: 946126-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR