4-Ethyl-4'-ethynyl-1,1'-biphenyl 97%

Supplier: Ambeed
Warning

A150781-25G A150781-5G
76783-368EA 81.69 USD
76783-368 76783-370
4-Ethyl-4'-ethynyl-1,1'-biphenyl 97%
4-Ethyl-4'-ethynyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₆H₁₄
MW: 206.28 g/mol
MDL Number: MFCD01333522
CAS Number: 477587-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR