(S)-N-Boc-3-methylmorpholine 97%

Supplier: Ambeed
Warning

Synonyms: (S)-N-Boc-3-Methylmorpholine

A150772-100MG A150772-1G A150772-250MG A150772-5G
76783-360EA 9.55 USD
76783-360 76783-362 76783-364 76783-366
(S)-N-Boc-3-methylmorpholine 97%
(S)-N-Boc-3-methylmorpholine

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MW: 201.2600 g/mol
MDL Number: MFCD12964052
CAS Number: 1022094-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR