3-Chloro-2-methoxybenzaldehyde 97%

Supplier: Ambeed
Warning

A150100-10G A150100-1G A150100-250MG A150100-25G A150100-5G
76783-344EA 152.77 USD
76783-344 76783-346 76783-348 76783-350 76783-352
3-Chloro-2-methoxybenzaldehyde 97%
3-Chloro-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇ClO₂
MW: 170.5930 g/mol
MDL Number: MFCD09999135
CAS Number: 223778-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR