4-Chlorothiazol-2-amine 95%

Supplier: Ambeed
Danger

A149930-100MG A149930-1G A149930-250MG
76783-328EA 90.18 USD
76783-328 76783-330 76783-332
4-Chlorothiazol-2-amine 95%
4-Chlorothiazol-2-amine

Precaución: for Research Use Only

Formula: C₃H₃ClN₂S
MW: 134.587 g/mol
MDL Number: MFCD09999189
CAS Number: 52107-46-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR