7-Methoxyquinoline 98%

Supplier: Ambeed
Warning

A149896-25G A149896-1G
76783-326EA 914.5 USD
76783-326 77667-134
7-Methoxyquinoline 98%
7-Methoxyquinoline

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MW: 159.19 g/mol
MDL Number: MFCD00870291
CAS Number: 4964-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR