1-Ethoxy-2,3-difluoro-4-(trans-4-propylcyclohexyl)benzene 98%

Supplier: Ambeed
Warning

A149305-1G A149305-5G
76783-288EA 12 USD
76783-288 76783-290
1-Ethoxy-2,3-difluoro-4-(trans-4-propylcyclohexyl)benzene 98%
1-Ethoxy-2,3-difluoro-4-(trans-4-propylcyclohexyl)benzene

Precaución: for Research Use Only

Formula: C₁₇H₂₄F₂O
MW: 282.37 g/mol
MDL Number: MFCD11053400
CAS Number: 174350-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR