5-Chloropyridazin-3-(2H)-one 97%

Supplier: Ambeed
Warning

A149271-10G A149271-1G A149271-250MG A149271-25G A149271-5G
76783-280EA 137 USD
76783-280 76783-282 76783-284 76783-286 77667-132
5-Chloropyridazin-3-(2H)-one 97%
5-Chloropyridazin-3-(2H)-one

Precaución: for Research Use Only

Formula: C4H3ClN2O
MDL Number: MFCD12198134
CAS Number: 660425-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR