Fmoc-3-(3-pyridyl)-D-alanine 99%

Supplier: Ambeed
Warning

Synonyms: Fmoc-β-(3-pyridyl)-D-Ala-OH, Fmoc-D-3-Pal-OH

A149266-100MG A149266-1G A149266-250MG A149266-5G
76783-274EA 8 USD
76783-274 76783-276 76783-278 77871-729
Fmoc-3-(3-pyridyl)-D-alanine 99%
Fmoc-3-(3-pyridyl)-D-alanine

Precaución: for Research Use Only

Formula: C₂₃H₂₀N₂O₄
MW: 388.423 g/mol
MDL Number: MFCD00080267
CAS Number: 142994-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR