2,4-Dimethoxy-4'-hydroxybenzophenone 97%

Supplier: Ambeed
Warning

Synonyms: (2,4-Dimethoxyphenyl)(4-hydroxyphenyl)methanone

A149101-100G A149101-25G A149101-5G
76783-268EA 253.56 USD
76783-268 76783-270 76783-272
2,4-Dimethoxy-4'-hydroxybenzophenone 97%
2,4-Dimethoxy-4'-hydroxybenzophenone

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₄
MW: 258.27 g/mol
MDL Number: MFCD00137850
CAS Number: 41351-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR