Trimethobenzamide hydrochloride 98%

Supplier: Ambeed
Danger

Synonyms: Trimethobenzamide HCl, N-[4-[2-(Dimethylamino)ethoxy]benzyl]-3,4,5-trimethoxybenzamide hydrochloride, N-(4-(2-(Dimethylamino)ethoxy)benzyl)-3,4,5-trimethoxybenzamide hydrochloride

A148362-100MG A148362-1MG
76783-256EA 5.31 USD
76783-256 77667-106
Trimethobenzamide hydrochloride 98%
Trimethobenzamide hydrochloride

Precaución: for Research Use Only

Formula: C₂₁H₂₈N₂O₅·HCl
MW: 424.9184 g/mol
MDL Number: MFCD00057999
CAS Number: 554-92-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR