3-Bromo-8-chloroimidazo[1,2-a]pyrazine 97%

Supplier: Ambeed
Warning

A148216-1G A148216-250MG A148216-25G A148216-5G
76783-250EA 16 USD
76783-250 76783-252 76783-254 77667-102
3-Bromo-8-chloroimidazo[1,2-a]pyrazine 97%
3-Bromo-8-chloroimidazo[1,2-a]pyrazine

Precaución: for Research Use Only

Formula: C6H3BrClN3
MDL Number: MFCD09909653
CAS Number: 143591-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR