(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-methoxyphenyl)propanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: Boc-Tyr(Me)-OH, N-Boc-O-methyl-L-tyrosine

A148207-100G A148207-1G A148207-25G A148207-5G
76783-244EA 154 USD
76783-244 76783-246 76783-248 77592-786
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-methoxyphenyl)propanoic acid 97%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-methoxyphenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₅
MW: 295.3 g/mol
MDL Number: MFCD00065603
CAS Number: 53267-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR