2-Nitro-3-(trifluoromethyl)aniline 97%

Supplier: Ambeed
Warning

A147894-1G A147894-250MG
76783-238EA 70 USD
76783-238 76783-240
2-Nitro-3-(trifluoromethyl)aniline 97%
2-Nitro-3-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₅F₃N₂O₂
MW: 206.122 g/mol
MDL Number: MFCD00042448
CAS Number: 386-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR