2-Acetyl-6-methoxynaphthalene 98%

Supplier: Ambeed
Warning

Synonyms: 6'-Methoxy-2'-acetonaphthone

A147348-100G A147348-25G A147348-10G A147348-500G
76783-224EA 17 USD
76783-224 77317-234 77567-036 77667-094
2-Acetyl-6-methoxynaphthalene 98%
2-Acetyl-6-methoxynaphthalene

Precaución: For research use only.

Formula: C₁₃H₁₂O₂
MDL Number: MFCD00021643
CAS Number: 3900-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR