3-Chloro-4-(trifluoromethyl)benzonitrile 96%

Supplier: Ambeed
Warning

A146970-10G A146970-1G A146970-25G A146970-5G
76783-216EA 35 USD
76783-216 76783-218 76783-220 76783-222
3-Chloro-4-(trifluoromethyl)benzonitrile 96%
3-Chloro-4-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₃ClF₃N
MDL Number: MFCD04972703
CAS Number: 1092460-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR