2,4-Dichloro-3-methyl benzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,4-Dichloro-3-methylbenzoic acid

A146953-100MG A146953-1G A146953-250MG A146953-5G
76783-208EA 8 USD
76783-208 76783-210 76783-212 76783-214
2,4-Dichloro-3-methyl benzoic acid 98%
2,4-Dichloro-3-methyl benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O₂
MDL Number: MFCD16659660
CAS Number: 83277-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR