3-Amino-6-bromoisoquinoline 97%

Supplier: Ambeed
Warning

A146544-100MG A146544-1G A146544-250MG A146544-5G A146544-25G
76783-174EA 16 USD
76783-174 76783-176 76783-178 76783-180 77667-090
3-Amino-6-bromoisoquinoline 97%
3-Amino-6-bromoisoquinoline

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MDL Number: MFCD11101002
CAS Number: 891785-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR