Boc-D-beta-Homophenylalanine 95%

Supplier: Ambeed
Warning

A145996-100MG A145996-1G A145996-250MG A145996-5G
76783-158EA 11 USD
76783-158 76783-160 76783-162 76783-164
Boc-D-beta-Homophenylalanine 95%
Boc-D-beta-Homophenylalanine

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₄
MDL Number: MFCD01076270
CAS Number: 101555-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR