Trientine dihydrochloride 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
Trientine HCl, N1,N1'-(Ethane-1,2-diyl)bis(ethane-1,2-diamine) dihydrochloride
A145937-250MG
A145937-25G
A145937-5G
A145937-1G
76783-152EA
24
USD
76783-152
76783-154
76783-156
77667-088
Trientine dihydrochloride 96%
Trientine dihydrochloride
Precaución: for Research Use Only
Formula:
C₆H₂₀Cl₂N₄ MW: 219.16 g/mol |
MDL Number:
MFCD00012889 CAS Number: 38260-01-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...