3,6-Dihydro-2H-pyran-4-carboxylicacid methyl ester 96%

Supplier: Ambeed
Warning

Synonyms: Methyl 3,6-dihydro-2H-pyran-4-carboxylate

A145911-100MG A145911-1G A145911-250MG A145911-5G
76783-144EA 51.99 USD
76783-144 76783-146 76783-148 76783-150
3,6-Dihydro-2H-pyran-4-carboxylicacid methyl ester 96%
3,6-Dihydro-2H-pyran-4-carboxylicacid methyl ester

Precaución: for Research Use Only

Formula: C₇H₁₀O₃
MW: 142.16 g/mol
MDL Number: MFCD11706987
CAS Number: 105772-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR