4'-Butyl-4-iodobiphenyl 97%

Supplier: Ambeed
Warning

Synonyms: 4-Butyl-4'-iodobiphenyl

A145899-1G A145899-250MG A145899-5G
76783-138EA 13.8 USD
76783-138 76783-140 76783-142
4'-Butyl-4-iodobiphenyl 97%
4'-Butyl-4-iodobiphenyl

Precaución: for Research Use Only

Formula: C₁₆H₁₇I
MW: 336.22 g/mol
MDL Number: MFCD16619281
CAS Number: 199982-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR