2-(Boc-amino)-6-chloropyridine 97%

Supplier: Ambeed
Warning

A145302-100G A145302-1G A145302-25G A145302-10G
76783-128EA 480 USD
76783-128 76783-130 76783-132 77871-712
2-(Boc-amino)-6-chloropyridine 97%
2-(Boc-amino)-6-chloropyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₃ClN₂O₂
MDL Number: MFCD09030821
CAS Number: 159603-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR