Tris(3-methylphenyl)phosphine 98%

Supplier: Ambeed
Warning

Synonyms: Tris(m-tolyl)phosphine, Tri-m-tolylphosphine

A144058-100G A144058-1G A144058-25G A144058-5G
76783-066EA 157.02 USD
76783-066 76783-068 76783-070 76783-072
Tris(3-methylphenyl)phosphine 98%
Tris(3-methylphenyl)phosphine

Precaución: for Research Use Only

Formula: C₂₁H₂₁P
MW: 304.37 g/mol
MDL Number: MFCD00008532
CAS Number: 6224-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR