5-Fluoro-m-cresol 98%

Supplier: Ambeed
Danger

Synonyms: 1-Fluoro-3-hydroxy-5-methylbenzene, 3-Fluoro-5-methylphenol

A143704-10G A143704-1G A143704-25G A143704-5G
76783-058EA 41 USD
76783-058 76783-060 76783-062 76783-064
5-Fluoro-m-cresol 98%
5-Fluoro-m-cresol

Precaución: for Research Use Only

Formula: C₇H₇FO
MW: 126.13
MDL Number: MFCD04115885
CAS Number: 216976-31-7
UN: 3265
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR