5-Acetylpyridin-2(1H)-one 97%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-5-acetylpyridine

A143514-100MG A143514-10G A143514-1G A143514-250MG A143514-5G A143514-25G
76783-032EA 6 USD
76783-032 76783-034 76783-036 76783-038 76783-040 77871-697
5-Acetylpyridin-2(1H)-one 97%
5-Acetylpyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₇H₇NO₂
MDL Number: MFCD10697705
CAS Number: 1124-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR