tert-Butyl methyl(2-oxoethyl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl methyll(2-oxoethyl)carbamate, Methyl(2-oxoethyl)carbamic acid tert-butyl ester, N-Boc-(methylamino)acetaldehyde, N-tert-Butoxycarbonyl-(methylamino)acetaldehyde

A141883-10G A141883-1G A141883-250MG A141883-25G A141883-5G A141883-100G
76782-972EA 36.08 USD
76782-972 76782-974 76782-976 76782-978 76782-980 77667-058
tert-Butyl methyl(2-oxoethyl)carbamate 95%
tert-Butyl methyl(2-oxoethyl)carbamate

Precaución: for Research Use Only

Formula: C₈H₁₅NO₃
MW: 173.21 g/mol
MDL Number: MFCD08064267
CAS Number: 123387-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR