4-Bromo-1H-benzoimidazole 98%

Supplier: Ambeed
Warning

A141721-250MG A141721-5G A141721-25G
76782-966EA 25.47 USD
76782-966 77566-852 77667-056
4-Bromo-1H-benzoimidazole 98%
4-Bromo-1H-benzoimidazole

Precaución: For research use only.

Formula: C₇H₅BrN₂
MW: 197.0320 g/mol
MDL Number: MFCD09754164
CAS Number: 83741-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR