2-Amino-5-bromopyrimidin-4-ol hydrobromide 95%

Supplier: Ambeed
Warning

A141671-100MG A141671-1G A141671-250MG A141671-5G
76782-952EA 49.87 USD
76782-952 76782-954 76782-956 76782-958
2-Amino-5-bromopyrimidin-4-ol hydrobromide 95%
2-Amino-5-bromopyrimidin-4-ol hydrobromide

Precaución: for Research Use Only

Formula: C₄H₅Br₂N₃O
MW: 270.9100 g/mol
MDL Number: MFCD12923111
CAS Number: 1215597-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR