Isoquinoline-3-carbaldehyde 97%

Supplier: Ambeed
Warning

A141131-100MG A141131-10G A141131-1G A141131-250MG A141131-5G
76782-918EA 11 USD
76782-918 76782-920 76782-922 76782-924 76782-926
Isoquinoline-3-carbaldehyde 97%
Isoquinoline-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₇NO
MDL Number: MFCD06227437
CAS Number: 5470-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR