6-Chloro-n,n-dimethylnicotinamide 97%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-N,N-dimethylnicotinamide

A140133-100MG A140133-1G A140133-250MG A140133-5G
76782-894EA 5.31 USD
76782-894 76782-896 76782-898 77566-806
6-Chloro-n,n-dimethylnicotinamide 97%
6-Chloro-n,n-dimethylnicotinamide

Precaución: For research use only.

Formula: C₈H₉ClN₂O
MW: 184.623 g/mol
MDL Number: MFCD08588286
CAS Number: 54864-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR