Methyl-2-amino-4,6-dichloronicotinate 95%

Supplier: Ambeed
Warning

A139587-100MG A139587-1G A139587-250MG A139587-5G
76782-872EA 6 USD
76782-872 76782-874 76782-876 76782-878
Methyl-2-amino-4,6-dichloronicotinate 95%
Methyl-2-amino-4,6-dichloronicotinate

Precaución: for Research Use Only

Formula: C₇H₆Cl₂N₂O₂
MDL Number: MFCD16660199
CAS Number: 1044872-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR