trans-4-(Cbz-amino)cyclohexan-1-ol 98%

Supplier: Ambeed
Warning

Synonyms: Benzyl (trans-4-hydroxycyclohexyl)carbamate

A139472-1G A139472-5G A139472-25G
76782-866EA 10 USD
76782-866 76782-868 76782-870 77667-016
trans-4-(Cbz-amino)cyclohexan-1-ol 98%
trans-4-(Cbz-amino)cyclohexan-1-ol

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₃
MW: 249.306 g/mol
MDL Number: MFCD00158431
CAS Number: 27489-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR