2-tert-Butylbenzoic acid 95%

Supplier: Ambeed
Warning

A139444-1G A139444-250MG A139444-5G A139444-100MG
76782-862EA 112.01 USD
76782-862 76782-864 77667-014 77871-672
2-tert-Butylbenzoic acid 95%
2-tert-Butylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MDL Number: MFCD09258829
CAS Number: 1077-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR