Z-D-Glu(OBzl)-OH 97%

Supplier: Ambeed
Warning

A139291-1G A139291-25G A139291-5G
76782-858EA 59 USD
76782-858 76782-860 77667-010
Z-D-Glu(OBzl)-OH 97%
Z-D-Glu(OBzl)-OH

Precaución: for Research Use Only

Formula: C₂₀H₂₁NO₆
MDL Number: MFCD00077014
CAS Number: 59486-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR