3-Bromo-4-fluoro-5-trifluoromethyl-phenylamine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-4-fluoro-5-(trifluoromethyl)aniline

A138810-100MG A138810-1G A138810-250MG A138810-5G
76782-826EA 9 USD
76782-826 76782-828 76782-830 76782-832
3-Bromo-4-fluoro-5-trifluoromethyl-phenylamine 98%
3-Bromo-4-fluoro-5-trifluoromethyl-phenylamine

Precaución: for Research Use Only

Formula: C₇H₄BrF₄N
MW: 258.011 g/mol
MDL Number: MFCD11520114
CAS Number: 1233026-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR