4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate 95%

Supplier: Ambeed
Warning

Synonyms: 4-Acetoxyphenylboronic acid pinacol ester, 2-(4-Acetoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl Acetate, 4-Acetoxyphenylboronic acid, pinacol ester, 4-Acetoxyphenylboronic acid Pinacol Ester

A138512-10G A138512-1G A138512-25G A138512-5G
76782-810EA 38 USD
76782-810 76782-812 76782-814 76782-816
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate 95%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate

Precaución: for Research Use Only

Formula: C₁₄H₁₉BO₄
MDL Number: MFCD03453060
CAS Number: 480424-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR