Verteporfin 98%

Supplier: Ambeed
Warning

Synonyms: BPD-MA, 4R,4aS)-18-Ethenyl-4,4a-dihydro-3,4-bis(methoxycarbonyl)-4a,8,14,19-tetramethyl-24H,26H-benzo[b]porphine-9,13-dipropanoic acid monomethyl ester, (4R,4aS)-rel-18-Ethenyl-4,4a-dihydro-3,4-bis(methoxycarbonyl)-4a,8,14,19-tetramethyl-24H,26H-Benzo[b]porphine-9,13-dipropanoic acid monomethyl ester

A138297-10MG A138297-1G A138297-1MG A138297-250MG A138297-5MG A138297-100MG A138297-50MG
76782-778EA 106 USD
76782-778 76782-780 76782-782 76782-784 76782-786 77600-010 77600-012
Verteporfin 98%
Verteporfin

Precaución: for Research Use Only

Formula: C₄₁H₄₂N₄O₈
MDL Number: MFCD28098202
CAS Number: 129497-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR