3-Fluoro-2-nitroanisole 98%

Supplier: Ambeed
Danger

A138158-10G A138158-1G A138158-250MG A138158-25G A138158-5G
76782-768EA 23 USD
76782-768 76782-770 76782-772 76782-774 76782-776
3-Fluoro-2-nitroanisole 98%
3-Fluoro-2-nitroanisole

Precaución: for Research Use Only

Formula: C₇H₆FNO₃
MDL Number: MFCD07368752
CAS Number: 641-49-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR