Tribromomethyl phenyl sulfone 97%

Supplier: Ambeed
Warning

Synonyms: ((Tribromomethyl)sulphonyl)benzene, Phenyl Tribromomethyl Sulphone

A137754-1G A137754-25G A137754-5G
76782-750EA 5.31 USD
76782-750 76782-752 77600-002
Tribromomethyl phenyl sulfone 97%
Tribromomethyl phenyl sulfone

Precaución: for Research Use Only

Formula: C₇H₅Br₃O₂S
MW: 392.89 g/mol
MDL Number: MFCD00060068
CAS Number: 17025-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR