trans-1-Ethoxy-2,3-difluoro-4-(4-pentylcyclohexyl)benzene 97%

Supplier: Ambeed
Warning

A137299-1G A137299-5G A137299-100G A137299-25G
76782-736EA 11 USD
76782-736 77666-976 77871-656 77871-657
trans-1-Ethoxy-2,3-difluoro-4-(4-pentylcyclohexyl)benzene 97%
trans-1-Ethoxy-2,3-difluoro-4-(4-pentylcyclohexyl)benzene

Precaución: for Research Use Only

Formula: C₁₉H₂₈F₂O
MDL Number: MFCD11053401
CAS Number: 124729-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR