14-Bromo-1-tetradecanol 95%

Supplier: Ambeed
Warning

Synonyms: 14-Bromotetradecan-1-ol

A137159-100MG A137159-1G A137159-250MG A137159-25G A137159-5G
76782-718EA 7 USD
76782-718 76782-720 76782-722 76782-724 76782-726
14-Bromo-1-tetradecanol 95%
14-Bromo-1-tetradecanol

Precaución: for Research Use Only

Formula: HO(CH₂)₁₃CH₂Br
MW: 293.29
MDL Number: MFCD01076225
CAS Number: 72995-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR